Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2065 in, 2.0000 in
Catalog No. 5000530746
Encompass
The Class ZZ Plus Plug Gage is a precision instrument designed for go inspection, setting micrometers, verniers, and other measuring devices. It is made from tool steel and features a 10 micro coating/finish, ensuring accuracy and durability. This plug gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro coating/finish.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-30746 Supplier MSC Supplier No. 88920657
The Class ZZ Plus Plug Gage is a precision instrument designed for go inspection, setting micrometers, verniers, and other measuring devices. It is made from tool steel and features a 10 micro coating/finish, ensuring accuracy and durability. This plug gage meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro coating/finish.
  • Meets ASME B89.1 standards.
  • Includes a certificate of accuracy with NIST traceability.