Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2140 in, 0.000200 in Tolerance
Catalog No. 5000528855
Encompass
This Class ZZ Plus Plug Gage is designed for precision inspection and calibration. Made from durable tool steel with a 10 micro finish, it ensures reliable performance for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability, adhering to ASME B89.1 standards.
  • Designed for go inspection applications
  • Used for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel for durability
  • Features a 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Manufactured in the United States
Catalog No. 50-005-28855 Supplier MSC Supplier No. 88921408
This Class ZZ Plus Plug Gage is designed for precision inspection and calibration. Made from durable tool steel with a 10 micro finish, it ensures reliable performance for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability, adhering to ASME B89.1 standards.
  • Designed for go inspection applications
  • Used for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel for durability
  • Features a 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Manufactured in the United States