Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2170 in Diameter, Tool Steel
Catalog No. 5000528854
Encompass
This Class ZZ Plus Plug Gage features a 0.2170 in diameter and a 2.0000 in overall length, constructed from durable tool steel. It is manufactured with a 0.000200 in tolerance and a 10 micro finish, meeting ASME B89.1 standards. The gage is ideal for go inspection and for setting micrometers, verniers, and other measuring devices, and includes a certificate of accuracy with NIST traceability.
  • 0.2170 in diameter
  • 2.0000 in overall length
  • Tool steel material
  • 0.000200 in tolerance
  • 10 micro finish
  • Meets ASME B89.1 standards
  • Suitable for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-28854 Supplier MSC Supplier No. 88921705
This Class ZZ Plus Plug Gage features a 0.2170 in diameter and a 2.0000 in overall length, constructed from durable tool steel. It is manufactured with a 0.000200 in tolerance and a 10 micro finish, meeting ASME B89.1 standards. The gage is ideal for go inspection and for setting micrometers, verniers, and other measuring devices, and includes a certificate of accuracy with NIST traceability.
  • 0.2170 in diameter
  • 2.0000 in overall length
  • Tool steel material
  • 0.000200 in tolerance
  • 10 micro finish
  • Meets ASME B89.1 standards
  • Suitable for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability