Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2325 in Dia, 0.0002 in Tolerance
Catalog No. 5000529027
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision tool made from durable tool steel, designed for accurate go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.2325 inch diameter with a tight 0.0002 inch tolerance and a 10 microfinish for reliable measurements. The overall length is 2.0000 inches.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-29027 Supplier MSC Supplier No. 88923255
The Vermont Gage Class ZZ Plus Plug Gage is a precision tool made from durable tool steel, designed for accurate go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.2325 inch diameter with a tight 0.0002 inch tolerance and a 10 microfinish for reliable measurements. The overall length is 2.0000 inches.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.