Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2330 in Dia, Tool Steel
Catalog No. 5000530537
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise "Go" inspection and setting measuring devices. This tool steel gage has a 0.2330 in diameter and an overall length of 2.0000 in, with a tolerance of 0.000200 in. It includes a Certificate of Accuracy with NIST Traceability for assured quality.
  • Designed for "Go" inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Adheres to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-30537 Supplier MSC Supplier No. 88923305
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise "Go" inspection and setting measuring devices. This tool steel gage has a 0.2330 in diameter and an overall length of 2.0000 in, with a tolerance of 0.000200 in. It includes a Certificate of Accuracy with NIST Traceability for assured quality.
  • Designed for "Go" inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Adheres to ASME B89.1 standards.
  • Features a 10 micro coating/finish.