Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2420 in Dia, Tool Steel
Catalog No. 5000515368
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement, ideal for "Go" inspection and setting micrometers, verniers, and other measuring devices. It features a 0.2420 inch diameter, 2.0000 inch overall length, and a tight 0.000200 inch tolerance. Constructed from durable tool steel with a 10 micro finish, it adheres to ASME B89.1 standards. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliability and precision.
  • Precision measurement for "Go" inspection.
  • Versatile use for setting measuring devices.
  • High accuracy with 0.000200 inch tolerance.
  • Durable tool steel construction.
  • Certificate of Accuracy with NIST traceability.
  • Compliant with ASME B89.1 standards.
Catalog No. 50-005-15368 Supplier MSC Supplier No. 88924204
This Class ZZ Plus Plug Gage is designed for precise measurement, ideal for "Go" inspection and setting micrometers, verniers, and other measuring devices. It features a 0.2420 inch diameter, 2.0000 inch overall length, and a tight 0.000200 inch tolerance. Constructed from durable tool steel with a 10 micro finish, it adheres to ASME B89.1 standards. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliability and precision.
  • Precision measurement for "Go" inspection.
  • Versatile use for setting measuring devices.
  • High accuracy with 0.000200 inch tolerance.
  • Durable tool steel construction.
  • Certificate of Accuracy with NIST traceability.
  • Compliant with ASME B89.1 standards.