Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2520 in Diameter, 2.0000 in Overall Length, 0.000200 in Tolerance
Catalog No. 5000531694
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 0.2520 in diameter and a 2.0000 in overall length, manufactured with a tight tolerance of 0.000200 in. The gage has a 10 micro-inch finish and adheres to ASME B89.1 standards, ensuring accuracy and reliability for go inspection, setting micrometers, verniers, and other measuring devices.
  • Precision measurement for various applications
  • Manufactured from durable tool steel
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro-inch finish
Catalog No. 50-005-31694 Supplier MSC Supplier No. 88925201
This Vermont Gage Class ZZ Plus Plug Gage is designed for high-precision measurement applications. It features a 0.2520 in diameter and a 2.0000 in overall length, manufactured with a tight tolerance of 0.000200 in. The gage has a 10 micro-inch finish and adheres to ASME B89.1 standards, ensuring accuracy and reliability for go inspection, setting micrometers, verniers, and other measuring devices.
  • Precision measurement for various applications
  • Manufactured from durable tool steel
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro-inch finish