Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2585 in Dia, 2.0000 in OAL
Catalog No. 5000531543
Encompass
This plug gage is designed for precise measurements and inspection tasks. It features a 0.2585 inch diameter and a 2.0000 inch overall length, making it suitable for various calibration and inspection applications.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 0.000200 inch tolerance
  • Made from tool steel
  • Has a 10 micro coating/finish
  • Manufactured in the United States
Catalog No. 50-005-31543 Supplier MSC Supplier No. 88925854
This plug gage is designed for precise measurements and inspection tasks. It features a 0.2585 inch diameter and a 2.0000 inch overall length, making it suitable for various calibration and inspection applications.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 0.000200 inch tolerance
  • Made from tool steel
  • Has a 10 micro coating/finish
  • Manufactured in the United States