Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2650 in Dia, 2.0000 in OAL, 0.000200 in Tolerance
Catalog No. 5000530752
Encompass
This Class ZZ Plus Plug Gage is designed for accurate "Go" inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.2650 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance and 10 Micro finish. Constructed from tool steel, this gage adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for "go" inspection and for setting micrometers, verniers, and other measuring devices
  • Manufactured from durable tool steel
  • Adheres to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Made in the United States
Catalog No. 50-005-30752 Supplier MSC Supplier No. 88926506
This Class ZZ Plus Plug Gage is designed for accurate "Go" inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.2650 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance and 10 Micro finish. Constructed from tool steel, this gage adheres to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for "go" inspection and for setting micrometers, verniers, and other measuring devices
  • Manufactured from durable tool steel
  • Adheres to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Made in the United States