Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2705" Diameter, 2.0000" OAL
Catalog No. 5000527384
Encompass
This plug gage measures 0.2705" in diameter with an overall length of 2.0000". Constructed from durable tool steel, it features a 10 microfinish and a tight tolerance of 0.000200". Designed for precision, it is suitable for 'Go' inspection and accurately setting various measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Conforms to ASME B89.1 standards.
Catalog No. 50-005-27384 Supplier MSC Supplier No. 88927058
This plug gage measures 0.2705" in diameter with an overall length of 2.0000". Constructed from durable tool steel, it features a 10 microfinish and a tight tolerance of 0.000200". Designed for precision, it is suitable for 'Go' inspection and accurately setting various measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for go inspection.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Conforms to ASME B89.1 standards.