Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.28 mm Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000529020
Encompass
The Class ZZ Plus Plug Gage is a precision measuring instrument crafted from durable tool steel, designed for accurate inspection and calibration tasks. With its specific dimensions and tight tolerances, this plug gage is an essential tool for quality control and precision measurement applications. Each gage comes with a Certificate of Accuracy, ensuring traceability to NIST standards.
  • High precision with 0.28 mm diameter and 0.00508 mm tolerance.
  • Durable tool steel construction.
  • Versatile for "Go" inspection and setting measuring instruments.
  • NIST traceable with Certificate of Accuracy.
  • Meets ASME B89.1 quality standards.
  • 10 micro coating/finish for precision and wear resistance.
  • Proudly manufactured in the United States.
  • Convenient 2.0000 inch overall length for ease of handling.
Catalog No. 50-005-29020 Supplier MSC Supplier No. 68091339
The Class ZZ Plus Plug Gage is a precision measuring instrument crafted from durable tool steel, designed for accurate inspection and calibration tasks. With its specific dimensions and tight tolerances, this plug gage is an essential tool for quality control and precision measurement applications. Each gage comes with a Certificate of Accuracy, ensuring traceability to NIST standards.
  • High precision with 0.28 mm diameter and 0.00508 mm tolerance.
  • Durable tool steel construction.
  • Versatile for "Go" inspection and setting measuring instruments.
  • NIST traceable with Certificate of Accuracy.
  • Meets ASME B89.1 quality standards.
  • 10 micro coating/finish for precision and wear resistance.
  • Proudly manufactured in the United States.
  • Convenient 2.0000 inch overall length for ease of handling.