Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2845 in Diameter, Tool Steel
Catalog No. 5000527418
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'go' inspection and for accurately setting micrometers, verniers, and other measuring devices. It features a specific diameter, overall length, and tight tolerance, ensuring reliable performance in various metrology applications.
  • Made from durable tool steel
  • Diameter of 0.2845 inch
  • Overall length of 2.0000 inch
  • Tolerance of 0.000200 inch
  • Ideal for 'go' inspection and setting measuring devices
  • Conforms to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
Catalog No. 50-005-27418 Supplier MSC Supplier No. 88928452
This Class ZZ Plus Plug Gage is designed for precise 'go' inspection and for accurately setting micrometers, verniers, and other measuring devices. It features a specific diameter, overall length, and tight tolerance, ensuring reliable performance in various metrology applications.
  • Made from durable tool steel
  • Diameter of 0.2845 inch
  • Overall length of 2.0000 inch
  • Tolerance of 0.000200 inch
  • Ideal for 'go' inspection and setting measuring devices
  • Conforms to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish