Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2880 in, 2.0000 in
Catalog No. 5000531544
Encompass
Vermont Gage Class ZZ Plus Plug Gages are precision tools used for go inspection and setting various measuring devices. Constructed from durable tool steel, these gages provide accurate measurements for applications requiring high precision.
  • For go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Features a tolerance of 0.000200 in.
  • Coating/finish of 10 micro.
  • Manufactured to ASME B89.1 standards.
Catalog No. 50-005-31544 Supplier MSC Supplier No. 88928809
Vermont Gage Class ZZ Plus Plug Gages are precision tools used for go inspection and setting various measuring devices. Constructed from durable tool steel, these gages provide accurate measurements for applications requiring high precision.
  • For go inspection.
  • Used to set micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Features a tolerance of 0.000200 in.
  • Coating/finish of 10 micro.
  • Manufactured to ASME B89.1 standards.