Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.2910 in Diameter, 2.0000 in Overall Length, Tool Steel
Catalog No. 5000531545
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.2910 inch diameter and a 2.0000 inch overall length, with a 10 microinch finish. This gage is ideal for go inspection and for setting various measuring devices.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Constructed from tool steel
  • Features a 10 microinch finish
Catalog No. 50-005-31545 Supplier MSC Supplier No. 88929104
The Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.2910 inch diameter and a 2.0000 inch overall length, with a 10 microinch finish. This gage is ideal for go inspection and for setting various measuring devices.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Constructed from tool steel
  • Features a 10 microinch finish