Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3005 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000510521
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. It features a specific diameter and overall length, manufactured from durable tool steel. It is suitable for 'Go' inspection and calibrating various measuring devices.
  • Diameter of 0.3005 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • 10 Micro finish
  • Conforms to ASME B89.1 standards
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Made from tool steel
Catalog No. 50-005-10521 Supplier MSC Supplier No. 88930052
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. It features a specific diameter and overall length, manufactured from durable tool steel. It is suitable for 'Go' inspection and calibrating various measuring devices.
  • Diameter of 0.3005 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • 10 Micro finish
  • Conforms to ASME B89.1 standards
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Made from tool steel