Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3020 in Dia, 2.0000 in OAL
Catalog No. 5000531565
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection applications. It features a 0.3020 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-31565 Supplier MSC Supplier No. 88930201
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection applications. It features a 0.3020 inch diameter and a 2.0000 inch overall length, crafted from durable tool steel.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.