Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3160 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000530859
Encompass
The Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a specific diameter and overall length, making it suitable for "Go" inspection and for setting various measuring devices. Constructed from durable tool steel, this gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Diameter of 0.3160 inches
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inches
  • 10 micro coating/finish
  • Constructed from tool steel
  • Suitable for "Go" inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-30859 Supplier MSC Supplier No. 88931605
The Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a specific diameter and overall length, making it suitable for "Go" inspection and for setting various measuring devices. Constructed from durable tool steel, this gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards.
  • Diameter of 0.3160 inches
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inches
  • 10 micro coating/finish
  • Constructed from tool steel
  • Suitable for "Go" inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards