Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3190 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000527389
Encompass
This economical Vermont Gage Class ZZ Plus Plug Gage offers precise measurements for various applications, including Go inspection and setting measuring devices. Made from durable tool steel, it adheres to ASME B89.1 standards and includes NIST traceability with a Certificate of Accuracy.
  • Economical pin gage for quick, accurate measurements
  • Measures hole sizes, distances, slot widths, and checks locations
  • Ideal for Go/NoGo gaging; sizes micrometers and snap gages
  • Designed for extended wear life
  • NIST traceable, includes Certificate of Accuracy
  • Made from tool steel (60/65 Rc)
  • Features 10 microinch finish or better
  • Round within 0.0001 inch
  • Class ZZ tolerance of 0.0002 inch
  • Overall length of 2.0000 inch
Catalog No. 50-005-27389 Supplier MSC Supplier No. 88931902
This economical Vermont Gage Class ZZ Plus Plug Gage offers precise measurements for various applications, including Go inspection and setting measuring devices. Made from durable tool steel, it adheres to ASME B89.1 standards and includes NIST traceability with a Certificate of Accuracy.
  • Economical pin gage for quick, accurate measurements
  • Measures hole sizes, distances, slot widths, and checks locations
  • Ideal for Go/NoGo gaging; sizes micrometers and snap gages
  • Designed for extended wear life
  • NIST traceable, includes Certificate of Accuracy
  • Made from tool steel (60/65 Rc)
  • Features 10 microinch finish or better
  • Round within 0.0001 inch
  • Class ZZ tolerance of 0.0002 inch
  • Overall length of 2.0000 inch