Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3200 in Dia, 2.0000 in OAL
Catalog No. 5000515812
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is engineered for precise 'Go' inspection tasks. With a 0.3200 inch diameter and a 2.0000 inch overall length, this gage is constructed from durable tool steel, ensuring reliability and longevity. It comes complete with a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards, making it suitable for setting various measuring devices.
  • Designed for 'Go' inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Constructed from durable tool steel.
  • Features a precision tolerance of 0.000200 "
  • Equipped with a 10 micro coating/finish.
Catalog No. 50-005-15812 Supplier MSC Supplier No. 88932009
The Vermont Gage Class ZZ Plus Plug Gage is engineered for precise 'Go' inspection tasks. With a 0.3200 inch diameter and a 2.0000 inch overall length, this gage is constructed from durable tool steel, ensuring reliability and longevity. It comes complete with a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards, making it suitable for setting various measuring devices.
  • Designed for 'Go' inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Constructed from durable tool steel.
  • Features a precision tolerance of 0.000200 "
  • Equipped with a 10 micro coating/finish.