Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3320 in, Tool Steel, NIST Traceable
Catalog No. 5000535748
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurements. It has a diameter of 0.3320 in, is constructed from durable tool steel, and includes a certificate of accuracy with NIST traceability.
  • Diameter of 0.3320 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Made from tool steel.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-35748 Supplier MSC Supplier No. 88933205
This Class ZZ Plus Plug Gage is designed for precision measurements. It has a diameter of 0.3320 in, is constructed from durable tool steel, and includes a certificate of accuracy with NIST traceability.
  • Diameter of 0.3320 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Made from tool steel.
  • Features a 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Suitable for Go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.