Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3370 in Diameter, 2.0000 in Overall Length, Tool Steel
Catalog No. 5000531720
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.3370 inch diameter and a 2.0000 inch overall length, adhering to ASME B89.1 standards for accuracy.
  • Class ZZ plus plug gage
  • 0.3370 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • Constructed from tool steel
  • 10 micro coating/finish
  • Adheres to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Used to set micrometers, verniers, and other measuring devices
Catalog No. 50-005-31720 Supplier MSC Supplier No. 88933700
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Made from durable tool steel, it features a 0.3370 inch diameter and a 2.0000 inch overall length, adhering to ASME B89.1 standards for accuracy.
  • Class ZZ plus plug gage
  • 0.3370 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • Constructed from tool steel
  • 10 micro coating/finish
  • Adheres to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Used to set micrometers, verniers, and other measuring devices