Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3435 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000531968
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications, including 'Go' inspection, setting micrometers, verniers, and other measuring devices. Constructed from hardened tool steel with a 10 microinch finish, it ensures accuracy with a 0.3435 inch diameter, 2 inch overall length, and a 0.0002 inch tolerance. Each gage is NIST traceable and includes a Certificate of Accuracy.
  • Application for go inspection, setting micrometers, verniers and other measuring devices.
  • Constructed from durable tool steel for longevity.
  • Features a 10 microinch finish or better for precision.
  • NIST traceability ensures accuracy.
  • Certificate of accuracy included with each gage.
  • Round within 0.0001 in.
  • Marked with size and direction of tolerance on gages over 0.0605 in.
Catalog No. 50-005-31968 Supplier MSC Supplier No. 88934351
This Class ZZ Plus Plug Gage is designed for precise measurement applications, including 'Go' inspection, setting micrometers, verniers, and other measuring devices. Constructed from hardened tool steel with a 10 microinch finish, it ensures accuracy with a 0.3435 inch diameter, 2 inch overall length, and a 0.0002 inch tolerance. Each gage is NIST traceable and includes a Certificate of Accuracy.
  • Application for go inspection, setting micrometers, verniers and other measuring devices.
  • Constructed from durable tool steel for longevity.
  • Features a 10 microinch finish or better for precision.
  • NIST traceability ensures accuracy.
  • Certificate of accuracy included with each gage.
  • Round within 0.0001 in.
  • Marked with size and direction of tolerance on gages over 0.0605 in.