Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3450 in, 10 Microinch Finish
Catalog No. 5000515143
Encompass
This Class ZZ Plus Plug Gage is designed for accurate measurement and inspection in various industrial applications. Made from durable tool steel, it provides reliable performance for setting measuring devices and Go inspection.
  • Tolerance of 0.0002 in for precise gaging
  • Overall length of 2.0000 in
  • Features a 10 microinch finish for smooth operation
  • Suitable for Go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Manufactured in the United States
Catalog No. 50-005-15143 Supplier MSC Supplier No. 88934500
This Class ZZ Plus Plug Gage is designed for accurate measurement and inspection in various industrial applications. Made from durable tool steel, it provides reliable performance for setting measuring devices and Go inspection.
  • Tolerance of 0.0002 in for precise gaging
  • Overall length of 2.0000 in
  • Features a 10 microinch finish for smooth operation
  • Suitable for Go inspection and setting measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Manufactured in the United States