Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3590 in Diameter, Tool Steel
Catalog No. 5000515965
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is engineered for high-precision measurement tasks. This gage, with a 0.3590 in diameter, is ideal for go inspection and calibrating various measuring instruments such as micrometers and verniers.
  • Diameter of 0.3590 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Manufactured from tool steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-15965 Supplier MSC Supplier No. 88935903
The Vermont Gage Class ZZ Plus Plug Gage is engineered for high-precision measurement tasks. This gage, with a 0.3590 in diameter, is ideal for go inspection and calibrating various measuring instruments such as micrometers and verniers.
  • Diameter of 0.3590 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Manufactured from tool steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices