This Class ZZ Plus Plug Gage is engineered for precise measurement and 'Go' inspection applications. Constructed from durable tool steel, it features a 10 microfinish and meets ASME B89.1 standards. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance and adherence to national standards.
- Engineered for precise measurement in 'Go' inspection applications.
- Features a 0.3705 inch diameter and 2.0000 inch overall length.
- Maintains a tight 0.0002 inch tolerance.
- Constructed from durable tool steel with a smooth 10 microfinish.
- Each gage is marked with its size and tolerance direction.
- Includes a Certificate of Accuracy with N.I.S.T. traceability.
- Conforms to ASME B89.1 standards for quality assurance.
- Ideal for setting micrometers, verniers, and other measuring devices.