Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.3990 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000515950
Encompass
This Class ZZ Plus Plug Gage, with a diameter of 0.3990 inches and an overall length of 2.0000 inches, is manufactured from durable tool steel. It is designed for precise 'go' inspection and can be used to set various measuring devices such as micrometers and verniers, adhering to ASME B89.1 standards.
  • Manufactured from tool steel.
  • Features a 10 microfinish.
  • Suitable for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Class ZZ tolerance for precise measurements.
Catalog No. 50-005-15950 Supplier MSC Supplier No. 88939905
This Class ZZ Plus Plug Gage, with a diameter of 0.3990 inches and an overall length of 2.0000 inches, is manufactured from durable tool steel. It is designed for precise 'go' inspection and can be used to set various measuring devices such as micrometers and verniers, adhering to ASME B89.1 standards.
  • Manufactured from tool steel.
  • Features a 10 microfinish.
  • Suitable for go inspection applications.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Class ZZ tolerance for precise measurements.