Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.4140 in, 2.0000 in, Tool Steel
Catalog No. 5000513831
Encompass
This Class ZZ Plus plug gage is engineered for precise measurement applications. Constructed from durable tool steel, it features a 0.4140 inch diameter, a 2.0000 inch overall length, and a 10 microinch finish, making it ideal for checking hole diameters and setting measuring devices.
  • Class ZZ tolerance of 0.0002 inch ensures high accuracy.
  • Made from tool steel for durability and long life.
  • Features a 2.0000 inch overall length.
  • Provides a 10 microinch finish or better for smooth operation.
  • Suitable for go inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Round within 0.0001 "
Catalog No. 50-005-13831 Supplier MSC Supplier No. 88941406
This Class ZZ Plus plug gage is engineered for precise measurement applications. Constructed from durable tool steel, it features a 0.4140 inch diameter, a 2.0000 inch overall length, and a 10 microinch finish, making it ideal for checking hole diameters and setting measuring devices.
  • Class ZZ tolerance of 0.0002 inch ensures high accuracy.
  • Made from tool steel for durability and long life.
  • Features a 2.0000 inch overall length.
  • Provides a 10 microinch finish or better for smooth operation.
  • Suitable for go inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Round within 0.0001 "