Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.4300 in Diameter, Tool Steel
Catalog No. 5000515734
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it ensures accuracy and reliability in various measurement applications.
  • For 'Go' inspection
  • Sets micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-15734 Supplier MSC Supplier No. 88943006
This Class ZZ Plus Plug Gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it ensures accuracy and reliability in various measurement applications.
  • For 'Go' inspection
  • Sets micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Conforms to ASME B89.1 standards