Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.50 mm Diameter, 2.0000 in Overall Length, Tool Steel
Catalog No. 5000535740
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.50 mm diameter and a 2.0000 in overall length, manufactured from durable tool steel with a black oxide finish. This gage offers a 0.00508 mm tolerance, making it suitable for demanding metrology applications.
  • Overall length of 2.0000 in
  • Tolerance of 0.00508 mm
  • Made from durable tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Features a black oxide coating/finish
  • Complies with ASME B89.1 standards
  • Diameter of measurement is 0.50 mm
  • Manufactured in the United States
Catalog No. 50-005-35740 Supplier MSC Supplier No. 56201411
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.50 mm diameter and a 2.0000 in overall length, manufactured from durable tool steel with a black oxide finish. This gage offers a 0.00508 mm tolerance, making it suitable for demanding metrology applications.
  • Overall length of 2.0000 in
  • Tolerance of 0.00508 mm
  • Made from durable tool steel
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Features a black oxide coating/finish
  • Complies with ASME B89.1 standards
  • Diameter of measurement is 0.50 mm
  • Manufactured in the United States