Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.5330 in Diameter, 0.000200 in Tolerance, Tool Steel
Catalog No. 5000515398
Encompass
This Class ZZ Plus Plug Gage is an economical, general-purpose pin gage for quick and accurate measurements. It is used for gaging hole sizes, distances between holes, slot widths, and Go/NoGo applications. Made from tool steel, it features a 0.5330 in diameter, 2.0000 in overall length, and a 0.000200 in tolerance.
  • Economical, general-purpose pin gage for quick and accurate measurements.
  • Measures hole sizes, distances between holes, slot widths, and performs Go/NoGo gaging.
  • Offers added wear life compared to closer tolerance gages.
  • Constructed from durable tool steel with a 10 microinch finish or better.
  • Round within 0.0001 in.
  • Includes a Certificate of Accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Gages over 0.0605 in are marked with size and direction of tolerance.
Catalog No. 50-005-15398 Supplier MSC Supplier No. 88953302
This Class ZZ Plus Plug Gage is an economical, general-purpose pin gage for quick and accurate measurements. It is used for gaging hole sizes, distances between holes, slot widths, and Go/NoGo applications. Made from tool steel, it features a 0.5330 in diameter, 2.0000 in overall length, and a 0.000200 in tolerance.
  • Economical, general-purpose pin gage for quick and accurate measurements.
  • Measures hole sizes, distances between holes, slot widths, and performs Go/NoGo gaging.
  • Offers added wear life compared to closer tolerance gages.
  • Constructed from durable tool steel with a 10 microinch finish or better.
  • Round within 0.0001 in.
  • Includes a Certificate of Accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Gages over 0.0605 in are marked with size and direction of tolerance.