Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.5430 in Diameter, Tool Steel
Catalog No. 5000530755
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. With a diameter of 0.5430 inches and an overall length of 2.0000 inches, it is suitable for use in various industrial and laboratory settings where accurate gaging is critical.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-30755 Supplier MSC Supplier No. 88954300
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. With a diameter of 0.5430 inches and an overall length of 2.0000 inches, it is suitable for use in various industrial and laboratory settings where accurate gaging is critical.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards