Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.5530 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000531461
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Featuring a 0.5530 inch diameter, a 2.0000 inch overall length, and a tight tolerance of 0.0002 inch, it is constructed from durable tool steel with a 10 micro coating/finish. This gage is ideal for go inspection, setting micrometers, verniers, and other measuring devices, and includes a certificate of accuracy with NIST traceability.
  • Diameter of measurement: 0.5530 in
  • Overall length: 2.0000 in
  • Tolerance: 0.0002 in
  • Constructed from tool steel
  • Features a 10 micro coating/finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Complies with ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31461 Supplier MSC Supplier No. 88955307
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. Featuring a 0.5530 inch diameter, a 2.0000 inch overall length, and a tight tolerance of 0.0002 inch, it is constructed from durable tool steel with a 10 micro coating/finish. This gage is ideal for go inspection, setting micrometers, verniers, and other measuring devices, and includes a certificate of accuracy with NIST traceability.
  • Diameter of measurement: 0.5530 in
  • Overall length: 2.0000 in
  • Tolerance: 0.0002 in
  • Constructed from tool steel
  • Features a 10 micro coating/finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Complies with ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability