Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.5620 in, 0.000200 in Tolerance
Catalog No. 5000531700
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.5620 inch diameter, a 2.0000 inch overall length, and a 0.000200 inch tolerance. It is suitable for Go inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Coating/finish: 10 micro
  • Meets ASME B89.1 standards
Catalog No. 50-005-31700 Supplier MSC Supplier No. 88956206
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.5620 inch diameter, a 2.0000 inch overall length, and a 0.000200 inch tolerance. It is suitable for Go inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from tool steel
  • Coating/finish: 10 micro
  • Meets ASME B89.1 standards