Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.6270 in Dia, 2.0000 in OAL
Catalog No. 5000513956
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is designed for accurate 'Go' inspection and for setting other measuring devices. Made from durable tool steel, this gage measures 0.6270 inch in diameter with an overall length of 2.0000 " It includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Class ZZ Plus plug gage
  • 0.6270 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Made of tool steel
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Designed for "Go" inspection and setting other measuring devices
Catalog No. 50-005-13956 Supplier MSC Supplier No. 88962709
The Vermont Gage Class ZZ Plus Plug Gage is designed for accurate 'Go' inspection and for setting other measuring devices. Made from durable tool steel, this gage measures 0.6270 inch in diameter with an overall length of 2.0000 " It includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Class ZZ Plus plug gage
  • 0.6270 inch diameter
  • 2.0000 inch overall length
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Made of tool steel
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Designed for "Go" inspection and setting other measuring devices