Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.6560 in Diameter, 0.000200 in Tolerance
Catalog No. 5000537181
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a 0.6560 inch diameter, a 2.0000 inch overall length, and a 0.000200 inch tolerance. The gage also has a 10 micro finish, ensuring accuracy and reliability for quality control and manufacturing processes.
  • Class ZZ plus plug gage
  • 0.6560 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • 10 micro finish
Catalog No. 50-005-37181 Supplier MSC Supplier No. 88965603
This Class ZZ Plus Plug Gage is designed for precision measurement applications. It features a 0.6560 inch diameter, a 2.0000 inch overall length, and a 0.000200 inch tolerance. The gage also has a 10 micro finish, ensuring accuracy and reliability for quality control and manufacturing processes.
  • Class ZZ plus plug gage
  • 0.6560 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • 10 micro finish