Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.75 mm Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000531175
Encompass
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a 0.75 mm diameter and a 2.0000 inch overall length.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for Go inspection and setting various measuring devices.
  • Conforms to ASME B89.1 standards.
  • Made from durable tool steel.
  • Features a 10 micro coating/finish.
  • Measures 0.75 mm in diameter.
  • Has an overall length of 2.0000 inches.
  • Provides a tolerance of 0.00508 mm.
Catalog No. 50-005-31175 Supplier MSC Supplier No. 68091800
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, it features a 0.75 mm diameter and a 2.0000 inch overall length.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for Go inspection and setting various measuring devices.
  • Conforms to ASME B89.1 standards.
  • Made from durable tool steel.
  • Features a 10 micro coating/finish.
  • Measures 0.75 mm in diameter.
  • Has an overall length of 2.0000 inches.
  • Provides a tolerance of 0.00508 mm.