Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.8300 in, 2.0000 in
Catalog No. 5000535789
Encompass
The Class ZZ Plus Plug Gage is a precision instrument designed for accurate measurement and inspection. Crafted from durable tool steel, it is ideal for Go inspection, setting micrometers, verniers, and calibrating other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 micro finish.
  • Suitable for a range of measurement applications.
Catalog No. 50-005-35789 Supplier MSC Supplier No. 88983002
The Class ZZ Plus Plug Gage is a precision instrument designed for accurate measurement and inspection. Crafted from durable tool steel, it is ideal for Go inspection, setting micrometers, verniers, and calibrating other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 micro finish.
  • Suitable for a range of measurement applications.