Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.8420 in Diameter, Tool Steel, ASME B89.1
Catalog No. 5000531976
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.8420 in diameter, 2.0000 in overall length, and is constructed from durable tool steel. It meets ASME B89.1 standards and is suitable for various metrology applications.
  • Diameter of 0.8420 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Made from tool steel for durability
  • 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States
Catalog No. 50-005-31976 Supplier MSC Supplier No. 88984208
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection. It features a 0.8420 in diameter, 2.0000 in overall length, and is constructed from durable tool steel. It meets ASME B89.1 standards and is suitable for various metrology applications.
  • Diameter of 0.8420 in
  • Overall length of 2.0000 in
  • Tolerance of 0.000200 in
  • Made from tool steel for durability
  • 10 micro coating/finish
  • Complies with ASME B89.1 standards
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States