Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.846 in, 0.0002 in Tolerance
Catalog No. 5000515572
Encompass
This plug gage is designed for precise measurement and calibration. Made from durable tool steel, it features a 0.846 inch diameter and a tolerance of 0.0002 inch, with a 10 microfinish for accuracy.
  • For go inspection
  • Set micrometers, verniers and other measuring devices
  • 10 microfinish
  • Certificate of accuracy with NIST traceability
  • ASME B89.1 standard
Catalog No. 50-005-15572 Supplier MSC Supplier No. 88984604
This plug gage is designed for precise measurement and calibration. Made from durable tool steel, it features a 0.846 inch diameter and a tolerance of 0.0002 inch, with a 10 microfinish for accuracy.
  • For go inspection
  • Set micrometers, verniers and other measuring devices
  • 10 microfinish
  • Certificate of accuracy with NIST traceability
  • ASME B89.1 standard