Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.8500 in Dia, 2.0000 in OAL, Tool Steel
Catalog No. 5000530756
Encompass
This Class ZZ Plus Plug Gage is designed for Go Inspection and for setting micrometers, verniers, and other measuring devices. Constructed from tool steel, it features a 10 micro finish.
  • Measures 0.8500 in diameter
  • Has an overall length of 2.0000 in
  • Features a tolerance of 0.000200 in
  • Made from tool steel
  • Includes a Certificate of Accuracy with NIST Traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro finish
  • Manufactured in the United States
Catalog No. 50-005-30756 Supplier MSC Supplier No. 88985007
This Class ZZ Plus Plug Gage is designed for Go Inspection and for setting micrometers, verniers, and other measuring devices. Constructed from tool steel, it features a 10 micro finish.
  • Measures 0.8500 in diameter
  • Has an overall length of 2.0000 in
  • Features a tolerance of 0.000200 in
  • Made from tool steel
  • Includes a Certificate of Accuracy with NIST Traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro finish
  • Manufactured in the United States