Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 0.8680 in, 0.0002 in
Catalog No. 5000514529
Encompass
The Class ZZ Plus Plug Gage is an economical general-purpose pin gage designed for high-accuracy go inspection and setting various measuring devices. It is made from tool steel, features a 0.8680 inch diameter with a 0.0002 inch tolerance, and has a 10 microinch finish. Each gage is 2 inches long and includes a Certificate of Accuracy with NIST traceability.
  • Designed for quick and accurate measurements.
  • Suitable for go inspection and setting measuring devices.
  • Manufactured from tool steel for durability.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 "
  • Includes a Certificate of Accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Marked with size and direction of tolerance for gages over 0.0605 "
Catalog No. 50-005-14529 Supplier MSC Supplier No. 88986807
The Class ZZ Plus Plug Gage is an economical general-purpose pin gage designed for high-accuracy go inspection and setting various measuring devices. It is made from tool steel, features a 0.8680 inch diameter with a 0.0002 inch tolerance, and has a 10 microinch finish. Each gage is 2 inches long and includes a Certificate of Accuracy with NIST traceability.
  • Designed for quick and accurate measurements.
  • Suitable for go inspection and setting measuring devices.
  • Manufactured from tool steel for durability.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 "
  • Includes a Certificate of Accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Marked with size and direction of tolerance for gages over 0.0605 "