Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.05 mm, 2.00 in OAL, Tool Steel
Catalog No. 5000527238
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go/no-go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this plug gage features a 1.05 mm diameter and an overall length of 2.00 in, with a tight tolerance of 0.00508 mm, ensuring high precision in measurement tasks.
  • Designed for accurate go inspection.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel for longevity.
  • Provides high precision measurements with a tight tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro-inch finish for smooth operation.
Catalog No. 50-005-27238 Supplier MSC Supplier No. 68092105
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go/no-go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this plug gage features a 1.05 mm diameter and an overall length of 2.00 in, with a tight tolerance of 0.00508 mm, ensuring high precision in measurement tasks.
  • Designed for accurate go inspection.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel for longevity.
  • Provides high precision measurements with a tight tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro-inch finish for smooth operation.