Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.16 mm, 0.00508 mm Tolerance
Catalog No. 5000527809
Encompass
This Class ZZ Plus Plug Gage is a precision measuring tool used for 'Go Inspection' and for accurately setting micrometers, verniers, and other measuring devices. It is constructed from durable tool steel with a black oxide finish, ensuring reliability and longevity for demanding laboratory and industrial applications.
  • Designed for Go Inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Diameter of 1.16 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Features a black oxide finish
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-27809 Supplier MSC Supplier No. 50749159
This Class ZZ Plus Plug Gage is a precision measuring tool used for 'Go Inspection' and for accurately setting micrometers, verniers, and other measuring devices. It is constructed from durable tool steel with a black oxide finish, ensuring reliability and longevity for demanding laboratory and industrial applications.
  • Designed for Go Inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Diameter of 1.16 mm
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Features a black oxide finish
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards