Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.2914 in, Go/No-Go
Catalog No. 5000514657
Encompass
This Class ZZ Plus Plug Gage, with a diameter of 1.2914 inches, is designed for precise measurement applications. It functions as a Go/No-Go gage, providing efficient and accurate dimensional inspection for various components.
  • Functions as a Go/No-Go gage
  • Features a 1.2914 in diameter
  • Class ZZ Plus for high accuracy
  • Handle sold separately
Catalog No. 50-005-14657 Supplier MSC Supplier No. 64758253
This Class ZZ Plus Plug Gage, with a diameter of 1.2914 inches, is designed for precise measurement applications. It functions as a Go/No-Go gage, providing efficient and accurate dimensional inspection for various components.
  • Functions as a Go/No-Go gage
  • Features a 1.2914 in diameter
  • Class ZZ Plus for high accuracy
  • Handle sold separately