Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.2990 inch, Class ZZ, Tool Steel
Catalog No. 5000530599
Encompass
Vermont Gage Class ZZ Plus Plug Gages provide economical, accurate measurements. These versatile pin gages are ideal for checking hole sizes, distances, slot widths, and for Go/NoGo applications. Made from durable tool steel (60/65 Rc), they are NIST traceable and include a Certificate of Accuracy. Gages feature a Class ZZ tolerance of .0002 inch (.005mm), are round within .0001 inch (.0025mm), and have a 10 microinch finish. Larger diameters, such as 1.2990 inch, are available as custom taperlock plug gages.
  • Economical and accurate general purpose pin gages
  • Versatile for hole sizing, distance checks, slot gaging, and Go/NoGo applications
  • Extended wear life
  • NIST traceable, includes certificate of accuracy
  • Class ZZ tolerance (.0002 inch or .005 mm)
  • Round within .0001 inch (.0025 mm) with 10 microinch finish
  • Gages over .0605 inch are marked with size and tolerance
Catalog No. 50-005-30599 Supplier MSC Supplier No. 64759012
Vermont Gage Class ZZ Plus Plug Gages provide economical, accurate measurements. These versatile pin gages are ideal for checking hole sizes, distances, slot widths, and for Go/NoGo applications. Made from durable tool steel (60/65 Rc), they are NIST traceable and include a Certificate of Accuracy. Gages feature a Class ZZ tolerance of .0002 inch (.005mm), are round within .0001 inch (.0025mm), and have a 10 microinch finish. Larger diameters, such as 1.2990 inch, are available as custom taperlock plug gages.
  • Economical and accurate general purpose pin gages
  • Versatile for hole sizing, distance checks, slot gaging, and Go/NoGo applications
  • Extended wear life
  • NIST traceable, includes certificate of accuracy
  • Class ZZ tolerance (.0002 inch or .005 mm)
  • Round within .0001 inch (.0025 mm) with 10 microinch finish
  • Gages over .0605 inch are marked with size and tolerance