Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.3117 in Diameter
Catalog No. 5000531526
Encompass
This Class ZZ Plus plug gage provides precise measurements for verifying hole sizes and tolerances. With a 1.3117 inch diameter, it is suitable for a wide range of inspection applications. Please note that the handle for this plug gage is sold separately.
  • Precise 1.3117 inch diameter
  • Class ZZ Plus tolerance for accuracy
  • Ideal for verifying hole sizes
  • Suitable for various inspection applications
Catalog No. 50-005-31526 Supplier MSC Supplier No. 64760283
This Class ZZ Plus plug gage provides precise measurements for verifying hole sizes and tolerances. With a 1.3117 inch diameter, it is suitable for a wide range of inspection applications. Please note that the handle for this plug gage is sold separately.
  • Precise 1.3117 inch diameter
  • Class ZZ Plus tolerance for accuracy
  • Ideal for verifying hole sizes
  • Suitable for various inspection applications