Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.75 mm, 2.0000 in
Catalog No. 5000531177
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. It features a 1.75 mm diameter and a 2.0000 inch overall length, manufactured from durable Tool Steel. This gage is supplied with a Certificate of Accuracy for NIST Traceability, ensuring reliable performance in various laboratory and industrial applications.
  • Class ZZ Plus tolerance for high precision.
  • 1.75 mm diameter for accurate measurements.
  • 2.0000 in overall length for versatility.
  • Constructed from Tool Steel for durability.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Adheres to ASME B89.1 standards.
  • Features a 10 Micro finish.
  • Manufactured in the United States.
Catalog No. 50-005-31177 Supplier MSC Supplier No. 68092808
This Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. It features a 1.75 mm diameter and a 2.0000 inch overall length, manufactured from durable Tool Steel. This gage is supplied with a Certificate of Accuracy for NIST Traceability, ensuring reliable performance in various laboratory and industrial applications.
  • Class ZZ Plus tolerance for high precision.
  • 1.75 mm diameter for accurate measurements.
  • 2.0000 in overall length for versatility.
  • Constructed from Tool Steel for durability.
  • Includes Certificate of Accuracy with NIST Traceability.
  • Suitable for "Go" inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Adheres to ASME B89.1 standards.
  • Features a 10 Micro finish.
  • Manufactured in the United States.