Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.81 mm, Tool Steel, ASME B89.1
Catalog No. 5000527373
Encompass
This cylindrical plug gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features a 1.81 mm diameter with a 0.00508 mm tolerance, crafted from durable tool steel, and finished to 10 micro. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements for quality control and calibration applications.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for 'Go' inspection, setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Meets ASME B89.1 standards.
  • Features a 10 micro finish.
  • Backed by a hassle-free guarantee.
Catalog No. 50-005-27373 Supplier MSC Supplier No. 68092865
This cylindrical plug gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features a 1.81 mm diameter with a 0.00508 mm tolerance, crafted from durable tool steel, and finished to 10 micro. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements for quality control and calibration applications.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for 'Go' inspection, setting micrometers, verniers, and other measuring devices.
  • Made from durable tool steel.
  • Meets ASME B89.1 standards.
  • Features a 10 micro finish.
  • Backed by a hassle-free guarantee.