Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 10.80 mm, 2.0000 in OAL, Tool Steel
Catalog No. 5000530586
Encompass
This Class ZZ Plus plug gage is designed for precise "Go Inspection" and for calibrating measuring devices like micrometers and verniers. It features a tolerance of 0.00508 mm and a 10 micro finish, ensuring high accuracy.
  • Designed for "Go inspection"
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 micro finish
Catalog No. 50-005-30586 Supplier MSC Supplier No. 68101856
This Class ZZ Plus plug gage is designed for precise "Go Inspection" and for calibrating measuring devices like micrometers and verniers. It features a tolerance of 0.00508 mm and a 10 micro finish, ensuring high accuracy.
  • Designed for "Go inspection"
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Manufactured from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 micro finish