Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 11.40 mm, Tool Steel
Catalog No. 5000530973
Encompass
The Vermont Gage Class ZZ Plus Plug Gage features an 11.40 mm diameter and an overall length of 2.0000 inches. It has a tolerance of 0.00508 mm and a 10 Micro coating/finish. This plug gage is made from Tool Steel and is designed for Go Inspection, as well as for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards. The product is manufactured in the UNITED STATES.
  • Precision 11.40 mm diameter for accurate measurements
  • Durable tool steel construction
  • 2.0000 in overall length
  • Ideal for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Made in the United States
Catalog No. 50-005-30973 Supplier MSC Supplier No. 68102458
The Vermont Gage Class ZZ Plus Plug Gage features an 11.40 mm diameter and an overall length of 2.0000 inches. It has a tolerance of 0.00508 mm and a 10 Micro coating/finish. This plug gage is made from Tool Steel and is designed for Go Inspection, as well as for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and adheres to ASME B89.1 standards. The product is manufactured in the UNITED STATES.
  • Precision 11.40 mm diameter for accurate measurements
  • Durable tool steel construction
  • 2.0000 in overall length
  • Ideal for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Features a 10 micro coating/finish
  • Made in the United States